79174BC
GDS PPMX Gel Cooling Block
BioControl, For use with GDS PickPen® PIPETMAX®
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41103037
Recommended Products
material
plastic rack
description
For use with GDS PICKPEN™ PIPETMAX™ System
manufacturer/tradename
BioControl Systems 79174
storage condition
room temperature
technique(s)
PCR: suitable
application(s)
food and beverages
pathogen testing
compatibility
BioControl GDS
storage temp.
room temp
General description
4°C, Pink/Purple, 96 Position Rack
Features and Benefits
- Maintain a sample temperature of less than 4°C for almost four hours (with lid on rack) at room temperature.
- The plastic changes from purple to pink as the temperature of the rack changes, enabling you to easily monitor the temperature of your samples.
Components
One box of 2 cooling blocks for PPMX
Testing Method
PCR
Legal Information
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service