Skip to Content

73068BC

GDS Laptop Power Cord US and Charger

BioControl Systems, for use with GDS laptop

Synonym(s):

GDS Laptop AC Adapter and Charger, GDS Laptop Charger and Power Cable for US, GDS Laptop Charging Cable US and Charger, GDS Laptop Power Brick US and Charger, GDS Laptop Power Cable US and Charger, GDS Laptop Power Connector US and Charger, GDS Laptop Power Cord and Charging Adapter (US), GDS Laptop Power Cord for US Outlet and Charger, GDS Laptop Power Supply US and Charger, Laptop Charger Cord

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122409
NACRES:
NC.81
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 1 unit

manufacturer/tradename

BioControl Systems

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS laptop

storage temp.

room temp

General description

The GDS Laptop is an essential part of the Assurance® GDS Rotor-Gene Q thermocycler. The GDS Laptop Power Cord is suitable for use in the NA region.

Assurance® GDS is an automated nucleic acid amplification system for the detection of pathogens in food, ingredients, and environmental samples.

Application

GDS Laptop Power Cord US and Charger are essential accessories to be used with the GDS Rotor-Gene Q laptop provided with the Assurance® GDS system.

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library