Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
NACRES:
NA.51
UNSPSC Code:
12352208
Form:
liquid
Solubility:
water: soluble at 20 °C
Storage temp.:
2-8°C
Quality Level
form
liquid
manufacturer/tradename
Novagen®
storage condition
OK to freeze, avoid repeated freeze/thaw cycles
solubility
water: soluble at 20 °C
shipped in
wet ice
storage temp.
2-8°C
Related Categories
General description
Genomic DNAs are highly purified preparations of high molecular weight genomic DNA from a variety of organisms. The isolation protocol minimizes shearing and removes contaminants that inhibit enzymatic procedures. Novagen genomic DNA is qualified for genomic analysis (including PCR) and library construction. At least 90% of the DNA is greater than 100 kbp in size, as analyzed by CHEF gel electrophoresis (>50 kbp for S. cerevisiae). No inhibition of restriction enzyme activity is observed, and the DNA is free of contaminating RNA and protein.
The DNA is supplied in 10 mM Tris-HCl, pH 8.0, 1 mM EDTA (TE). Pipet with a wide-borepipet tip to minimize shearing.
The DNA is supplied in 10 mM Tris-HCl, pH 8.0, 1 mM EDTA (TE). Pipet with a wide-borepipet tip to minimize shearing.
High molecular weight, high purity DNA for any application
Legal Information
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 69239-3 | 04055977274646 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service