Skip to Content
Merck
All Photos(1)

Key Documents

676481

Sigma-Aldrich

VEGFR Tyrosine Kinase Inhibitor II

The VEGFR Tyrosine Kinase Inhibitor II, also referenced under CAS 269390-69-4, controls the biological activity of VEGFR Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

VEGFR Tyrosine Kinase Inhibitor II, N-(4-Chlorophenyl)-2-[(pyridin-4-ylmethyl)amino]benzamide, VEGF Receptor Tyrosine Kinase Inhibitor II, VEGFR2 Kinase Inhibitor XVII, VEGFR1 Kinase Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16ClN3O
CAS Number:
Molecular Weight:
337.80
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 5 mg/mL
methanol: soluble

shipped in

wet ice

storage temp.

−20°C

SMILES string

Clc1ccc(cc1)NC(=O)c2c(cccc2)NCc3ccncc3

InChI

1S/C19H16ClN3O/c20-15-5-7-16(8-6-15)23-19(24)17-3-1-2-4-18(17)22-13-14-9-11-21-12-10-14/h1-12,22H,13H2,(H,23,24)

InChI key

GGPZCOONYBPZEW-UHFFFAOYSA-N

General description

A potent, cell-permeable, reversible, and ATP-competitive inhibitor of the kinase activities of KDR, Flt-1 and c-Kit (IC50 = 20 nM, 180 nM and 240 nM, respectively), and minimally inhibit c-Src and EGF-R activities (IC50 = 7.0 µM and 7.3 µM). Also displays antiangiogenic and antitumor properties. Does not inhibit the activities of CDK-1, c-Met, IGF-1R or PKA (IC50 >10 µM).
A pyridinyl-anthranilamide compound that displays both antiangiogenic and antitumor properties. A potent, cell-permeable, reversible, and ATP-competitive inhibitor of the kinase activities of KDR, Flt-1 and c-Kit (IC50 = 20 nM, 180 nM and 240 nM, respectively), and minimally inhibit c-Src and EGF-R activities (IC50 = 7 µM and 7.3 µM). Further, inactive towards the inhibition of CDK-1, c-Met, IGF-1R and PKA (IC50 >10 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Kinase activities of KDR, Flt-1 and c-Kit
Product competes with ATP.
Reversible: yes
Target IC50: 20 nM, 180 nM and 240 nM against kinase activities of KDR, Flt-1 and c-Kit, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Furet, P., et al. 2003. Bioorg. Med. Chem. Lett.13, 2967.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shu-Yun Li et al.
Biology of reproduction, 105(4), 958-975 (2021-05-20)
Testis differentiation is initiated when Sry in pre-Sertoli cells directs the gonad toward a male-specific fate. Sertoli cells are essential for testis development, but cell types within the interstitial compartment, such as immune and endothelial cells, are also critical for
Perivascular cells support folliculogenesis in the developing ovary.
Li, et al.
Proceedings of the National Academy of Sciences of the USA, 119, e2213026119-e2213026119 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service