5871
Thiamine, Hydrochloride
Cofactor required for oxidation of carbohydrates and for synthesis of ribose.
Synonym(s):
Thiamine, Hydrochloride, Vitamin B₁
About This Item
Recommended Products
Quality Level
description
RTECS - XI7350000
Merck USA index - 14, 9295
Assay
≥98% (by assay)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
solubility
water: 100 mg/mL
shipped in
ambient
storage temp.
15-25°C
SMILES string
[s]1c[n+](c(c1CCO)C)Cc2c[n+H]c(nc2N)C.[Cl-].[Cl-]
InChI
1S/C12H17N4OS.2ClH/c1-8-11(3-4-17)18-7-16(8)6-10-5-14-9(2)15-12(10)13;;/h5,7,17H,3-4,6H2,1-2H3,(H2,13,14,15);2*1H/q+1;;/p-1
InChI key
DPJRMOMPQZCRJU-UHFFFAOYSA-M
General description
Warning
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service