Skip to Content
Merck

565758

β-Secretase Substrate IV, Fluorogenic

≥95% (HPLC), lyophilized solid, β-secretase substrate, Calbiochem®

Synonym(s):

β-Secretase Substrate IV, Fluorogenic, FS-1, H-RE(EDANS)EVNLDAEFK(DABCYL)R-OH, Memapsin 2 Substrate I, Fluorogenic

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C91H129N25O25S
CAS Number:
Molecular Weight:
2005.22
MDL number:
NACRES:
NA.77
UNSPSC Code:
12352202
Assay:
≥95% (HPLC)
Form:
lyophilized solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

β-Secretase Substrate IV, Fluorogenic, A highly sensitive fluorescence resonance energy transfer (FRET) peptide substrate, based on the EVNLDAEF sequence derived from the β-secretase site of the Swedish mutation of Amyloid Precursor Protein (APP).

Quality Segment

assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

red

solubility

DMSO: 5 mg/mL

fluorescence

λex ~350 nm, λem ~490 nm

shipped in

ambient

storage temp.

−20°C

General description

A highly sensitive fluorescence resonance energy transfer (FRET) peptide substrate (excitation max.: ~350 nm; emission max.: ~490), based on the GVNLDAEF sequence derived from the β-secretase site of the Swedish mutation of Amyloid Precursor Protein. Hydrolysis of the substrate at Leu-Asp bond by β-secretase results in fluorescence enhancement. Reported Kcat/km values (min-1/µM) for pro-mempasin 2 and its proteolytically activated species Leu28p-mempasin 2 are 0.044 and 0.040, respectively (at pH 4.5, 37°C).

Biochem/physiol Actions

Product does not compete with ATP.
Reversible: no
Cell permeable: no
Primary Target
β-secretase substrate

Packaging

Packaged under inert gas

Physical form

Supplied as a trifluoroacetate salt.

Preparation Note

Following reconstitution, aliquot and freeze at -20°C. Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Ermolieff, J., et al. 2000. Biochemistry 39, 12450.
H-Arg-Glu(EDANS)Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-Lys(DABCYL)Arg-OH

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
565758-1MG04055977267709