Skip to Content
Merck

5.55362

DiluCult Booster Pump

DiluCult, DiluCult 2, Accessory for faster and efficient functioning of the dilutors

Synonym(s):

Support pump

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
41122409
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

manufacturer/tradename

DiluCult

application(s)

food and beverages
sample preparation

compatibility

DiluCult 2, DiluCult

storage temp.

2-30°C

General description

Accurate microbiology results are highly reliant upon the quality of your sample preparation. Streamline your sample enrichment with our DILUCULT gravimetric dilutors. These automated gravimetric dilutor instruments have two integrated pumps and are designed to facilitate work in a microbiological safety cabinet. The booster pump enables 2X faster flowrate for faster and efficient functioning of the dilutors.

Legal Information

DILUCULT is a trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library