Skip to Content
Merck

5202

L-Phenylalanine

Synonym(s):

L-Phenylalanine, Phe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H11NO2
CAS Number:
Molecular Weight:
165.19
UNSPSC Code:
12352209
NACRES:
NA.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

description

RTECS - AY7535000
Merck USA index - 14, 7271

Quality Level

assay

≥98.5% dry basis (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

pH

5.4-6.0 (20 °C, 10 g/L)

mp

275-283 °C

solubility

water: soluble

density

1.34 g/cm3 at 20 °C

cation traces

heavy metals (as Pb): ≤20 ppm

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

15-25°C

SMILES string

[N+H3]C(Cc1ccccc1)C(=O)[O-]

InChI

1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)

InChI key

COLNVLDHVKWLRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at https://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 year at -20°C.

Other Notes

For a complete product listing of our ICP and AAS range, including technical information and example certificates, please visit ICP & AAS standards website.

Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
B43205-1G04061826736586
5202-100GM04055977271911

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service