Skip to Content
Merck
All Photos(1)

Key Documents

5.09265

Sigma-Aldrich

CX-5461

≥97% (HPLC), powder, RNA polymerase inhibitor, Calbiochem®

Synonym(s):

RNA Polymerase I Inhibitor II, CX-5461, Pol I Inhibitor II, CX5461, 2-(4-Methyl-1,4-diazepan-1-yl)-N-((5-methylpyrazin-2-yl)methyl)-5-oxo-5H-benzo[4,5]thiazolo[3,2-a][1,8]naphthyridine-6-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H27N7O2S
CAS Number:
Molecular Weight:
513.61
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

RNA Polymerase I Inhibitor II, CX-5461,

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 0.5 mg/mL
dilute acetic acid: 10 mg/mL

storage temp.

2-8°C

SMILES string

[s]1c2[n](c4nc(ccc4[c](c2C(=O)NCc6ncc(nc6)C)=O)N5CCN(CCC5)C)c3c1cccc3

InChI

1S/C27H27N7O2S/c1-17-14-29-18(15-28-17)16-30-26(36)23-24(35)19-8-9-22(33-11-5-10-32(2)12-13-33)31-25(19)34-20-6-3-4-7-21(20)37-27(23)34/h3-4,6-9,14-15H,5,10-13,16H2,1-2H3,(H,30,36)

InChI key

XGPBJCHFROADCK-UHFFFAOYSA-N

General description

A cell-permeable benzothiazole compound that selectively inhibits RNA polymerase I-mediated pre-rRNA transcription (IC50 = 142 nM in HCT-116), but not Pol II-mediated c-myc transcription, by preventing TAF SL1 and rDNA association). Effectively inhibits Pol I-dependent proliferation among 39 cancer cell lines (IC50<1 µM) in vitro and suppresses A375 (50 mg/kg/d p.o.) and MIA PaCa-2 (50 mg/kg/3d p.o.) tumor expansion in vivo.
A cell-permeable benzothiazole compound that selectively inhibits RNA polymerase I-mediated pre-rRNA transcription (IC50 = 142, 113, 54, and 74 nM in HCT-116, A375, MIA PaCa-2, and BJ-hTert cells, respectively), but not Pol II-mediated c-myc transcription, by preventing TAF (TATA Binding Protein-/TBP-Associated Factor) SL1 and rDNA association, effectively inhibiting Pol I-dependent proliferation among 39 cancer cell lines (IC50<1 µM) in vitro. While Pol I inhibition by CX-5461 is shown to result in p53-dependent apoptosis in cancer cells of hematopoietic origin, cell death by CX-5461 treatment in solid tumors-derived A375 and MIA PaCa-2 cultures are primarily due to autophagy and senescence, but not apoptosis, induction. Shown to suppress A375 (50 mg/kg/d p.o.) and MIA PaCa-2 (50 mg/kg/3d p.o.) tumor expansion, as well as selectively induce p53-mediated cell death of Eμ-Myc lymphoma, but not normal B cells, in mice (50 mg/kg p.o.) in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Pol I multi-protein complex
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Haddach, M., et al. 2012. ACS Med. Chem. Lett.3, 602.
Bywater, M.J., et al. 2012. Cancer Cell22, 51.
Drygin, D., et al. 2011. Cancer Res71, 1418.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service