Skip to Content
Merck

480010-M

NDSB-256

A non-detergent sulfobetaine. Reported to prevent protein aggregation and facilitate the renaturation of chemically and thermally denatured proteins.

Synonym(s):

NDSB-256, Dimethylbenzylammonium Propane Sulfonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H19NO3S
CAS Number:
Molecular Weight:
257.35
UNSPSC Code:
12161902
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic)

color

white

solubility

water: 200 mg/mL

storage temp.

10-30°C

SMILES string

[S](=O)(=O)([O-])CCC[N+](Cc1ccccc1)(C)C

InChI

1S/C12H19NO3S/c1-13(2,9-6-10-17(14,15)16)11-12-7-4-3-5-8-12/h3-5,7-8H,6,9-11H2,1-2H3

InChI key

MEJASPJNLSQOAG-UHFFFAOYSA-N

General description

A member of the class of nondetergent sulfobetaines. Unlike CHAPS and other zwittergents, NDSBs are nondenaturing even at 1 M concentrations. NDSBs have been used to improve the efficiency of solubilization of membrane proteins, to isolate nuclear proteins, to chromatographically purify halophilic proteins in low ionic strength buffers, and to crystallize proteins. They are zwitterionic over a wide pH range and do not absorb significantly in the near UV range. Reported to prevent protein aggregation and facilitate the renaturation of chemically and thermally denatured proteins. NDSBs can be easily removed by dialysis. At 1 M, NDSB-256 will restore 30% of the enzymatic activity of denatured egg white lysozyme; at 800 mM it will restore 16% of the enzymatic activity of denatured β-galactosidase.
A non-detergent sulfobetaine. Reported to prevent protein aggregation and facilitate the renaturation of chemically and thermally denatured proteins. Zwitterionic over a wide pH range. Easily removed by dialysis. At 1 M, NDSB-256 will restore 30% of the enzymatic activity of denatured egg white lysozyme; at 800 mM it will restore 16% of the enzymatic activity of denatured β-galactosidase.

Preparation Note

Following reconstitution, store at room temperature (15-30°C). Stock solutions are stable for up to 3 months at room temperature.

Other Notes

Goldberg, M.E., et al. 1996. Folding & Design1, 21.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library