Skip to Content
Merck
All Photos(1)

Documents

386032

Sigma-Aldrich

Anti-Hsp70 Mouse mAb (C92F3A-5)

liquid, clone C92F3A-5, Calbiochem®

Synonym(s):

Anti-Heat Shock Protein 70

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

C92F3A-5, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity (predicted by homology)

all

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSPA1A(3303)

General description

Anti-Hsp70, mouse monoclonal, clone C92F3A-5, recognizes the ~70 kDa Hsp70. Does not cross-react with Hsc70. It is validated for use in ELISA, FC, WB, ICC, IP & IHC (frozen and paraffin sections).
Mouse monoclonal antibody purified by ion-exchange chromatography. Recognizes the ~70 kDa Hsp70 protein.
Recognizes the ~70 kDa Hsp70 protein. Does not cross-react with Hsc70.

Immunogen

Hsp70 from HeLa cells
Human

Application

ELISA (see comments)

Flow Cytometry (see comments)

Frozen Sections (5 µg/ml)

Immunoblotting (1 µg/ml)

Immunocytochemistry (5 µg/ml)

Immunoprecipitation (see comments)

Paraffin Sections (5 µg/ml)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 50% glycerol, pH 7.2.

Reconstitution

Following iniital thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
L929 cells, Human colon cancer tissue

Other Notes

Does not cross-react with Hsc70. Because Hsp70 is not expressed constitutively in most cells, this antibody is suitable for ascertaining whether a stress-response has occurred in the cell. Increased levels of Hsp70 expression occurs following stress even in cells that constitutively express Hsp70. This antibody has also been reported to work for ELISA, flow cytometry, and immunoprecipitation. Variables associated with assay conditions will dictate the optimal working dilution.
Hang, H., and Fox, M.H. 1995. Cytometry19, 119.
Kilgore, J.L., et al. 1994. J. Appl. Physiol.76, 589.
Heufelder, A.E., et al. 1992. J. Clin. Endocrinol. Metab.74, 724.
Gower, D.J., et al. 1989. J. Neurosurg.70, 605.
Milarski, K., et al. 1989.J. Cell Biol.108, 413.
Vass, K., et al. 1988. Acta Neuropathologica77, 128.
Welch, W.J., and Suhan, J.P. 1986. J. Cell Biol.103, 2035.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aigang Lu et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 22(2), 183-195 (2002-02-02)
Estradiol reduces brain injury from many diseases, including stroke and trauma. To investigate the molecular mechanisms of this protection, the effects of 17-beta-estradiol on heat shock protein (HSP) expression were studied in normal male and female rats and in male
Daniel Laubitz et al.
Experimental physiology, 91(5), 867-875 (2006-05-27)
The heat shock response is associated with the intracellular expression of a number of highly conserved heat shock proteins (Hsps). According to their molecular size, Hsps have been divided into several groups, which are strongly conserved and show high homology
Rosario Barranco et al.
International journal of legal medicine, 133(5), 1461-1467 (2019-06-22)
The diagnosis of drowning is one of the most difficult in forensic medicine. The aim of this study was to analyze pulmonary tissue reactions in death by drowning. In particular, we focused on the immunohistochemical expression of P-selectin, SP-A, HSP70
Anne K Voss et al.
The EMBO journal, 25(15), 3652-3663 (2006-07-22)
The mechanisms regulating the size of the cerebral cortex are poorly understood. Here, we demonstrate that the Rap1 guanine nucleotide exchange factor, C3G (Grf2, Rapgef1), controls the size of the cerebral precursor population. Mice lacking C3G show overproliferation of the
James H Ahn et al.
Assay and drug development technologies, 9(3), 236-246 (2010-12-08)
Heat shock protein 70 (Hsp70) is a chaperone protein that helps protect against cellular stress, a function that may be co-opted to fight human diseases. In particular, the upregulation of Hsp70 can suppress the neurotoxicity of misfolded proteins, suggesting possible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service