Skip to Content
Merck

382149

Histone Deacetylase Inhibitor III

The Histone Deacetylase Inhibitor III, also referenced under CAS 251456-60-7, controls the biological activity of Histone Deacetylase. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Histone Deacetylase Inhibitor III, M344, 4-Dimethylamino- N-(6-hydroxycarbamoylhexyl)benzamide, N-Hydroxy-7-(4-dimethylaminobenzoyl)aminoheptanamide, M344, 4-Dimethylamino-N-(6-hydroxycarbamoylhexyl)benzamide, N-Hydroxy-7-(4-dimethylaminobenzoyl)aminoheptanamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H25N3O3
CAS Number:
Molecular Weight:
307.39
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white

solubility

DMSO: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N(O)C(=O)CCCCCCNC(=O)c1ccc(cc1)N(C)C

InChI

1S/C16H25N3O3/c1-19(2)14-10-8-13(9-11-14)16(21)17-12-6-4-3-5-7-15(20)18-22/h8-11,22H,3-7,12H2,1-2H3,(H,17,21)(H,18,20)

InChI key

MXWDSZWTBOCWBK-UHFFFAOYSA-N

General description

A cell-permeable amide analog of Trichostatin A (Cat. No. 647925) that potently inhibits histone deacetylases (IC50 = 40 nM for rat liver HDAC and IC50 = 100 nM for maize HD). Induces differentiation and inhibits proliferation (~2 µM) of murine erythroleukemia cells.
A cell-permeable, amide analog of Trichostatin A (Cat. No. 647925) that potently inhibits histone deacetylase (IC50 = 40 nM for rat liver HDAC and IC50 = 100 nM for maize HDAC). Induces differentiation and inhibits proliferation (~2 µM) of murine erythroleukemia cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Rat liver HDAC
Product does not compete with ATP.
Reversible: no
Target IC50: 40 nM for rat liver HDAC

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Ramiszewski, S.W., et al. 2002. J. Med. Chem.45, 753.
Jung, M., et al. 1999. J. Med. Chem.42, 4669.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library