309795
DIDS disodium salt
≥85% (HPLC), solid, anion transport inhibitor, Calbiochem®
Synonym(s):
DIDS, Disodium Salt, 4,4ʹ-Diisothiocyanostilbene-2,2ʹ-disulfonic Acid, 2Na
About This Item
Recommended Products
Product Name
DIDS, Disodium Salt, Binds covalently and irreversibly to the outer surface of human erythrocyte membranes.
Quality Level
Assay
≥85% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated
protect from light
color
yellow
solubility
water: 0.5%
shipped in
ambient
storage temp.
−20°C
SMILES string
[S](=O)(=O)(O)c1c(ccc(c1)N=C=S)\C=C\c2c(cc(cc2)N=C=S)[S](=O)(=O)O
InChI
1S/C16H10N2O6S4/c19-27(20,21)15-7-13(17-9-25)5-3-11(15)1-2-12-4-6-14(18-10-26)8-16(12)28(22,23)24/h1-8H,(H,19,20,21)(H,22,23,24)/b2-1+
InChI key
YSCNMFDFYJUPEF-OWOJBTEDSA-N
General description
Biochem/physiol Actions
Cl- uptake
Warning
Other Notes
Horie, S., et al. 1993. Res. Commun. Chem. Pathol. Pharmacol. 79, 117.
Mayinger, P., and Meyer, D.I. 1993. EMBO J. 12, 659.
Catantchik, Z.I., and Rothstein, A. 1974. J. Membr. Biol.15, 207.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service