Skip to Content
Merck
All Photos(1)

Key Documents

197345

Sigma-Aldrich

BCL6 Inhibitor, 79-6

The BCL6 Inhibitor, 79-6 controls the biological activity of BCL6. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

BCL6 Inhibitor, 79-6, CID5721353, 2-((5Z)-5-(5-Bromo-2-oxo-1H-indol-3-ylidene)-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)butanedioic acid, B-Cell Lymphoma 6 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H9BrN2O6S2
Molecular Weight:
457.28
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark red

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable oxindole compound that selectively inhibits the transcriptional repression activity of BCL6 (by ~70% at 50 µM in 293T-based reporter assays), but not that of three other BTB/POZ (bric-a-brac tramtrack broad complex/Poxvirus zinc finger) family members, Kaiso, HIC1, and PLZF, by direct BCL6 binding (Kd = 138 µM) via the BTB domain and blocking corepressor BBD- (BCL6 binding domain) mediated BCL6 interaction (IC50 = 212 µM & Ki = 147 µM in competitive binding against SMRT-BBD peptide), resulting in upregulation of BCL6 target genes in BCL-dependent DLBCL (diffuse large B cell lymphoma), but not BCL-independent Toledo and OCI-Ly4 cell lines. Shown to exhibit antiproliferative activity against DLBCL cell lines (IC50 from 24 to 936 µM) as well as primary human DLBCL cells in vitro and suppress BCL-dependent tumor growth in mice (<40% of control tumor size on day 9; 50 mg/kg/day via i.p.) in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Cerchietti, L.C., et al. 2010. Cancer Cell17, 400.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maryamsadat Seyedsadr et al.
The Journal of clinical investigation, 134(15) (2024-08-01)
Inflammatory neuropathies, which include chronic inflammatory demyelinating polyneuropathy (CIDP) and Guillain Barré syndrome (GBS), result from autoimmune destruction of the PNS and are characterized by progressive weakness and sensory loss. CD4+ T cells play a key role in the autoimmune
Pamela L Graney et al.
Advanced functional materials, 30(48) (2021-03-12)
Antibiotic-resistant bacteria are a major global health threat that continues to rise due to a lack of effective vaccines. Of concern are Klebsiella pneumoniae that fail to induce in vivo germinal center B cell responses, which facilitate antibody production to
Choong-Hyun Koh et al.
Cancer immunology research, 8(5), 698-709 (2020-03-04)
Although treatment with the glucocorticoid-induced tumor necrosis factor receptor-related protein (GITR) agonistic antibody (DTA-1) has shown antitumor activity in various tumor models, the underlying mechanism is not fully understood. Here, we demonstrate that interleukin (IL)-21-producing follicular helper T (Tfh) cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service