Skip to Content
Merck

1.93227

Potassium fluoride

for analysis EMPARTA® ACS

Synonym(s):

Potassium fluoride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
KF
CAS Number:
Molecular Weight:
58.10
UNSPSC Code:
12352302
MDL number:
Grade:
ACS reagent
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

storage temp.

room temp

SMILES string

[K+].[F-]

InChI

1S/FH.K/h1H;/q;+1/p-1

InChI key

NROKBHXJSPEDAR-UHFFFAOYSA-M

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Potassium fluoride


pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library