Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
assay
≥95% (SDS-PAGE)
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, avoid repeated freeze/thaw cycles
shipped in
wet ice
storage temp.
−20°C
General description
Packaging
Physical form
Preparation Note
Other Notes
Hasse, R., et al. 1998. Electrophoresis 9, 569.
Catapano, A.L., et al. 1978. J. Lipid Res. 19, 1047.
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description Native Apolipoprotein C-III from human plasma found in VLDL and chylomicrons. Involved in triglyceride uptake by cells. Inhibits lipoprotein lipase and uptake of lipoprotein remnants by the liver. | description Native apolipoprotein E from human plasma. A major component of VLDL and IDL. Serves as a major cholesterol carrier. Essential for the normal catabolism of triglyceride-rich lipoprotein constituents. | description Native apolipoprotein B from human plasma. It is a dominant protein constituent of LDL and VLDL. Acts as a ligand for LDL receptor, directing clearance of LDL from plasma to the liver. | description lyophilized powder |
| assay ≥95% (SDS-PAGE) | assay ≥95% (SDS-PAGE) | assay ≥95% (SDS-PAGE) | assay - |
| form liquid | form liquid | form solid | form lyophilized powder |
| storage condition avoid repeated freeze/thaw cycles, OK to freeze | storage condition OK to freeze, avoid repeated freeze/thaw cycles | storage condition OK to freeze | storage condition - |
| shipped in wet ice | shipped in wet ice | shipped in ambient | shipped in - |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



