Skip to Content
Merck
All Photos(2)

Key Documents

1.46394

Millipore

Sabouraud Dextrose Agar

Chloramphenicol, bottle capacity 250 mL, bottle filling volume 200 mL , filling volume, closure type, blue screw cap and 3 loci, box of 6 bottles

Synonym(s):

SDA with Chloramphenicol, SDA with Cm, Sabouraud Dextrose Agar with neutralizer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Agency

EP 2.6.13
JP 4.05
USP 62

Quality Level

description

For determination of the total count of yeasts and molds

sterility

sterile

form

gelatinous solid

shelf life

9 mo.

feature

closure type blue screw cap and 3 loci
ready-to-use

composition

Chloramphenicol

packaging

box of 6 bottles

technique(s)

direct inoculation: suitable
sterile filtration: suitable

bottle capacity

250 mL

bottle filling volume

200 mL , filling volume

capacity

250 mL

pH

5.6

application(s)

cosmetics
food and beverages
microbiology
pharmaceutical

storage temp.

2-25°C

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Sabouraud Dextrose Agar (SDA) is a complex medium for cultivation and isolation of yeasts and molds. The high concentration of dextrose and low pH promotes the growth of yeast and molds while inhibiting bacterial growth. The media also promotes the formation of spores (conidia and sporangia) and pigments of yeasts and molds. The presence of chloramphenicol further inhibits the growth of the accompanying bacterial flora of heavily contaminated material, due to its wide spectrum.

Application

Sabouraud Dextrose Agar (SDA) with Chloramphenicol is designed for the determination of the total count of yeasts and molds. The media is used for the microbiological examination of non-sterile products.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Our media are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Storage and Stability

  • The product can be used for sampling until the expiry date if stored upright, protected from light, and properly sealed at +2 °C to +25 °C.
  • The testing procedures described on the CoA can be started up to the expiry date printed on the label.

Other Notes

Agar media should not be melted a second time after solidifying.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service