Skip to Content
Merck

1.44161

Reference Standard Endotoxin

EDQM endotoxin standard for Monocyte Activation Test

Synonym(s):

Endotoxin standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116130
Feature:
performs endotoxin standard curve in the MAT
Packaging:
pkg of 1 vial
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

EP Chapter 2.6.30

Quality Level

form

solid

feature

performs endotoxin standard curve in the MAT

packaging

pkg of 1 vial

manufacturer/tradename

PyroMAT®

application(s)

pharmaceutical
pyrogen testing

compatibility

configured for PyroDetect System, configured for PyroMAT® System

shipped in

dry ice

storage temp.

−20°C

suitability

pyrogenic bacteria

General description

The Reference Standard Endotoxin (RSE) is the endotoxin standard recommended by the European Pharmacopeia. The endotoxin activates the TLR4 receptor of monocytes.

Application

The Reference Standard Endotoxin is used to perform the endotoxin standard curve required to quantify pyrogens with the Monocyte Activation Test during pyrogen detection.

Features and Benefits

Officially valid for the uses prescribed in the European Pharmacopoeia

Preparation Note

Reconstituted RSE at -40°C or below

Other Notes

Endotoxin Standard, lyophilized - 1μg
PyroDetect Kit (1.44154)
PyroDetect Cryoblood (1.44155)
PyroMAT® Kit (PYR0MATKIT)
PyroMAT® Cells (PYR0MATCELLS)
NEP Control of HKSA (MATHKSA)
NEP Control of Flagellin (MATFLAGELLIN)

Legal Information

PYROMAT is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
144161000104027269258573