12-123
PKC Substrate Peptide
PKC Substrate Peptide primarily used in Kinase Assays.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.42
Recommended Products
Quality Level
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable (kinase)
NCBI accession no.
UniProt accession no.
shipped in
wet ice
Gene Information
human ... PRKCA(5578)
Biochem/physiol Actions
Protein Target: PKC
Target Sub-Family: AGC
Quality
routinely evaluated in an enzyme assay using PKC (14-115)
Physical form
Lyophilized powder
Storage and Stability
2 years at 4°C
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
L M Neri et al.
The Journal of biological chemistry, 273(45), 29738-29744 (1998-10-29)
It is well established that an independent inositide cycle is present within the nucleus, where it is involved in the control of cell proliferation and differentiation. Previous results have shown that when Swiss 3T3 cells are treated with insulin-like growth
Y L Siow et al.
The Journal of biological chemistry, 272(12), 7586-7594 (1997-03-21)
The 78-kDa protein kinase Mekk1 plays an important role in the stress response pathway that involves the activation of downstream kinases Sek1 and stress-activated protein kinase/c-Jun NH2-terminal kinase. Conserved serine and threonine residues located between the kinase subdomains VII and
I Yasuda et al.
Biochemical and biophysical research communications, 166(3), 1220-1227 (1990-02-14)
Among various phosphate acceptor proteins and peptides so far tested, a synthetic peptide having the sequence surrounding Ser(8) of myelin basic protein, Gln-Lys-Arg-Pro-Ser(8)-Gln-Arg-Ser-Lys-Tyr-Leu, (MBP4-14), is the most specific and convenient substrate which can be used for selective assay of protein
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service