Skip to Content
Merck

1.20087

Millipore

Eshmuno® HCX Resin

Eshmuno® HCX

Synonym(s):

Eshmuno® HCX

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101511

ligand

carboxymethyl
phenyl
sulfonate group

Quality Level

sterility

sterile (Caustic Stable)

product line

Eshmuno®

form

resin

specific analyte(s)

MAbs

parameter

1000 cm/hr flow rate
8 bar max. pressure

matrix active group

polymer, polyvinyl ether

capacity

50 mg binding capacity (pIgG/ml of resin)

transition temp

flash point 35 °C (calculated)

application(s)

recombinant protein
vaccine development

separation technique

mixed mode

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

The mixed-mode cation exchanger Eshmuno® HCX media was specifically designed to capture recombinant proteins at higher salt concentrations directly in typical ion exchange and flow-through applications

Features and Benefits

Eshmuno® HCX chromatography resin enables:

  • Greater capacity at high-salt concentrations
  • Superior productivity
  • Outstanding selectivity
  • Rigid base beads for easy packing
  • Excellent pressure-flow behavior
  • Strong chemical stability

Packaging

  • 1.20087.0100: Eshmuno® HCX Resin 100ml
  • 1.20087.0010: Eshmuno® HCX Resin 10ml
  • 1.20087.0500: Eshmuno® HCX Resin 500ml
  • 1.20087.5000: Eshmuno® HCX Resin 5L

Plastic bottle

Analysis Note

Appearance: Milky cream coloured turbid suspension, free from coarse mechanical impurities.
Microscopic evaluation: Spherical particles, no agglomerates, no fines.
Extractable matter (water): ≤ 0.08 %
Ionic capacity: 170 - 300 µeq/ml
Performance test (chymotrypsinogen): 75 - 95 mS/cm
Performance test (cytochrom C): 55 - 74 mS/cm
Pressure drop(column: ID=1.6 cm, L=10 cm at 5 ml/min): ≤ 1.0 bar
Particle size (50-120 µm): ≥ 80 %
Particle size (d50): 75 - 95 µm
Cerium: ≤ 30 µg/g
Endotoxins: ≤ 1.00 EU/ml
TAMC (total aerobic microbial count): ≤ 100 CFU/ml
TYMC (total yeast and mould count): ≤ 100 CFU/ml

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service