Skip to Content
Merck

1.10293

MUELLER-HINTON broth

for testing the sensitivity of clinically important pathogens

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106213
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.2-7.6 (21 g/L in H2O, after autoclaving)

solubility

21 g/L

bulk density

700 kg/m3

application(s)

clinical
microbiology

storage temp.

15-25°C

suitability

Bacillus spp., Enterococcus spp., Escherichia coli, Listeria spp., Pseudomonas spp., Staphylococcus spp., Streptococcus spp.

General description

for testing the sensitivity of clinically important pathogens

Analysis Note

Appearance (clearness)clearAppearance (colour)yellowishAppearance (5 days; room temperature)clearpH-value (25 °C)7.2 - 7.6Growth (Escherichia coli ATCC 25922 (WDCM 00013))good to very goodGrowth (Staphylococcus aureus ATCC 25923 (WDCM 00034))good to very goodGrowth (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025))good to very goodGrowth (Enterococcus faecalis ATCC 33186)good to very goodGrowth (Streptococcus pyogenes ATCC 12344)good to very goodGrowth (Streptococcus pneumoniae ATCC 6301)fair to goodGrowth (Listeria monocytogenes ATCC 19118)fair to goodGrowth (Bacillus subtilis ATCC 6633 (WDCM 00003))good to very goodTest of antagonistspasses testIncubation: 24 hrs.; 35 °C; aerobic.Ca (Calcium)Mg (Magnesium)

Other Notes

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under MUELLER-HINTON broth


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library