Skip to Content
Merck
All Photos(2)

Key Documents

1.07054

Supelco

n-Pentane

for analysis EMPARTA®

Synonym(s):

Pentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-692-4
bp:
35-36 °C (lit.)

vapor density

2.48 (vs air)

Quality Level

product line

EMPARTA®

form

liquid

autoignition temp.

500 °F

potency

>2000 mg/kg LD50, oral (Rat)

expl. lim.

~8.3 %

dilution

(for routine analytical applications)

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

transition temp

flash point -48 °C (refers to pure substance)

solubility

0.4 g/L

density

0.626 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service