Skip to Content
Merck
All Photos(2)

Key Documents

1.05588

Supelco

Sodium hydroxide solution

min. 10% (1.11), for analysis EMSURE®

Synonym(s):

Sodium hydroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305
NACRES:
NA.21

Product Name

Sodium hydroxide solution min. 10% (1.11), for analysis EMSURE®

vapor pressure

3 mmHg ( 37 °C)

Quality Level

product line

EMSURE®

form

liquid

concentration

10.0-10.5% NaOH

impurities

≤0.0001% Total nitrogen (N)
≤0.0005% Al (Aluminium)
≤0.0005% Chloride (Cl)
≤0.0005% Fe (Iron)
≤0.0005% Heavy metals (as Pb)
≤0.0005% Phosphate (PO4)
≤0.0005% Sulphate (SO?)
≤0.001% Silicate (SiO2)
≤1% Carbonate (as Na2CO3)

pH

13.8 (20 °C in H2O)

density

1.11 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Efficient Removal of Aqueous Heavy Metals: A study leveraging sodium hydroxide solution (minimum 10%) in the preparation of Na/N co-doped seaweed biochar composites for the removal of Pb(II) and Cu(II) from water. This research underscores the role of sodium hydroxide in environmental remediation technologies (Fu et al., 2024).

  • Acute Esophagitis Model: Sodium hydroxide solution is utilized in the treatment of acute esophagitis in a rat model, highlighting its potential therapeutic applications in biomedical research (Gurlek et al., 2024).

  • Dye Adsorption: The solution is used in the synthesis of kaolin-supported nickel oxide composites for catalytic oxidative degradation of methylene blue dye, serving as a pivotal study in water treatment and chemical engineering (Mohammed et al., 2024).

  • Chitosan Production: Research on the production of chitosan from shrimp shell utilizing sodium hydroxide solution followed by subcritical water hydrolysis, illustrating its critical role in advancing material science and sustainable production methods (Matouri et al., 2024).

Analysis Note

Assay (acidimetric): 10.0 - 10.5 %
Carbonate (as Na₂CO₃): ≤ 1 %
Chloride (Cl): ≤ 0.0005 %
Phosphate (PO₄): ≤ 0.0005 %
Silicate (SiO₂): ≤ 0.001 %
Sulphate (SO₄): ≤ 0.0005 %
Total nitrogen (N): ≤ 0.0001 %
Heavy metals (as Pb): ≤ 0.0005 %
Al (Aluminium): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Photometric determination with pyridylazoresorcinol (PAR) subsequent to acid extraction

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service