Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
form
solid
Quality Segment
composition
Dye, ≥90% spectrophotometric assay
pH
2.4 (20 °C, 10 g/L in H2O)
mp
159 °C
solubility
110 g/L
bulk density
400‑500 kg/m3
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
2-30°C
InChI
1S/2C23H25N2.3C2H2O4/c2*1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4;3*3-1(4)2(5)6/h2*5-17H,1-4H3;3*(H,3,4)(H,5,6)/q2*+1;;;/p-2
InChI key
CNYGFPPAGUCRIC-UHFFFAOYSA-L
General description
Application
Analysis Note
Identity
- oxalate:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (water): 616 - 620 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.003 g/l; water): 1730 - 1960
TLC-Test: passes test
Loss on drying (50 °C / <1 hPa): ≤ 7%
Suitability for microscopy: passes test
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| form solid | form powder or crystals | form crystalline | form - |
| application(s) diagnostic assay manufacturing | application(s) diagnostic assay manufacturing | application(s) diagnostic assay manufacturing | application(s) cleaning products |
| solubility 110 g/L | solubility H2O: 1 mg/mL, blue to very deep blue | solubility methanol: 0.01 g/10 mL, blue to very deep blue | solubility - |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 100 |
| storage temp. 2-30°C | storage temp. room temp | storage temp. room temp | storage temp. - |
| composition Dye, ≥90% spectrophotometric assay | composition Dye content, ≥90% | composition - | composition - |
signalword
Danger
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2
Storage Class
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.





