Skip to Content
Merck
All Photos(1)

Key Documents

1.02780

Supelco

Copper(II) sulfate pentahydrate

very fine crystals EMPLURA®

Synonym(s):

Copper(II) sulfate pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
249.68
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-847-6
NACRES:
NA.21
form:
solid
solubility:
317 g/L
Pricing and availability is not currently available.

Quality Level

product line

EMPLURA®

Assay

≥98.0% (iodometric)

form

solid

potency

960 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

pH

3.5-4.5 (20 °C, 50 g/L in H2O)

mp

147 °C

solubility

317 g/L

density

2.284 g/cm3 at 20 °C

cation traces

Fe: ≤0.5%

storage temp.

2-30°C

SMILES string

[Cu+2].[S](=O)(=O)([O-])[O-].O.O.O.O.O

InChI

1S/Cu.H2O4S.5H2O/c;1-5(2,3)4;;;;;/h;(H2,1,2,3,4);5*1H2/q+2;;;;;;/p-2

InChI key

JZCCFEFSEZPSOG-UHFFFAOYSA-L

Related Categories

General description

very fine crystals EMPLURA®

Analysis Note

Assay (iodometric)≥ 98.0 %Fe (Iron)≤ 0.5 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Can we autoclave the solution of Copper II sulfate, for experiments requiring sterile solutions??

    1 answer
    1. Autoclaving Copper(II) sulfate solution is not advisable for sterilization. The high temperatures and pressures can cause the decomposition of Copper(II) sulfate, potentially releasing harmful fumes and altering the solution's chemical composition. Instead, consider using alternative sterilization methods, such as filtration or chemical sterilization, that do not involve high temperatures.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service