Skip to Content
Merck
All Photos(1)

Key Documents

1.01772

Supelco

Petroleum benzine boiling range 40-60°C

for gas chromatography ECD and FID SupraSolv®

Synonym(s):

Petroleum ether, Petroleum spirit, Petroleum ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.03
technique(s):
gas chromatography (GC): suitable
bp:
40-60 °C/1013 hPa
vapor pressure:
350 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

350 hPa ( 20 °C)

Quality Level

product line

SupraSolv®

form

liquid

autoignition temp.

250 °C (DIN 51794)

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.8-7.4 % (v/v)

technique(s)

gas chromatography (GC): suitable

kinematic viscosity

0.45 cSt(20 °C)

bp

40-60 °C/1013 hPa

mp

<-80 °C

transition temp

flash point <-21 °C

density

0.645-0.665 g/cm3 at 15 °C

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

SupraSolv ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service