Skip to Content
Merck

1.01090

Aluminium oxide 60 G neutral (type E)

for thinlayer chromatography

Synonym(s):

Aluminium oxides-PLC, Aluminium oxides-TLC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.21
UNSPSC Code:
41115711
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

aluminum oxide matrix, plastic support

form

powder

feature

binder Gypsum, fluorescent indicator: no

packaging

pkg of 2.5 kg

technique(s)

thin layer chromatography (TLC): suitable

particle size

5-40 μm

pore size

60 Å medium pore diameter

pH

7.6 (20 °C, 100 g/L in H2O, slurry)

mp

2050 °C

density

4.0 g/cm3 at 20 °C

bulk density

500 kg/m3

storage temp.

2-30°C

General description

Standardized sorbents ensure reliable results every time. Our broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40 µm: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).

Application


  • Matrix Solid-Phase Dispersion Coupled with HPLC-UV for Simultaneous Extraction, Purification and Determination of Six Lignans in Schisandra chinensis Fruits.: This study presents an innovative analytical method involving the use of aluminium oxide 60 G neutral as part of the matrix solid-phase dispersion process, highlighting its efficiency in complex sample preparation and purification processes in analytical chemistry (Song et al., 2022).



  • Alumina physically loaded by thiosemicarbazide for selective preconcentration of mercury(II) ion from natural water samples.: Explores the use of aluminium oxide in enhancing the specificity and efficiency of mercury ion capture from environmental samples, crucial for environmental monitoring and pollution studies (Ahmed, 2008).


  • Heterogeneous Fenton oxidation of Direct Black G in dye effluent using functional kaolin-supported nanoscale zero iron.: Demonstrates the catalytic prowess of aluminium oxide in advanced oxidation processes, serving as a critical component in the treatment of industrial waste, relevant to chemists working with catalytic methods for pollution remediation (Liu et al., 2014).



Analysis Note

Assay (CaSO₄·0.5 H₂O): 7 - 10 %
pH-value: 7.0 - 8.5
d 10 (Laser diffraction, size distribution): ≤ 5 µm
d 50 (laser diffraction, size distribution): ≤ 15 µm
d 90 (Laser diffraction, size distribution): ≤ 30 µm


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library