Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.41
UNSPSC Code:
41116124
Pricing and availability is not currently available.
Quality Segment
IVD
for in vitro diagnostic use
dilution
(for the detection of microcalcification)
application(s)
clinical testing
diagnostic assay manufacturing
hematology
histology
storage temp.
15-25°C
General description
The Silver plating kit according to von Kossa - for detection of microcalcification, is used to visualize calcium deposits in histological tissue specimens. It contains two ready-to-use reagents that are needed for the silver plating. A counterstaining can be performed with, e. g. Nuclear fast red staining solution, optionally, which is not contained in the kit. Silver ions of the silver nitrate solution react with the carbonate and phosphate ions of the calcium in the stored calcium deposits and displace the calcium ions. These silver ions are reduced to metallic silver by exposure to strong light, and this silver is evaluated by microscopy.
The package provides 300 stainings. This product is registered as IVD and CE marked.
For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.
The package provides 300 stainings. This product is registered as IVD and CE marked.
For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.
Application
For detection of calcium deposits in paraffin sections
Analysis Note
Suitability for microscopypasses testCalciumbrown to blackNucleiredCollagenredBackgroundred
signalword
Danger
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Met. Corr. 1 - Repr. 1B - Skin Irrit. 2
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


