Skip to Content
Merck
All Photos(1)

Key Documents

LM1304

Avanti

17:0-14:1 PS

Avanti Research - A Croda Brand LM1304, methanol solution

Synonym(s):

1-heptadecanoyl-2-(9Z-tetradecenoyl)-sn-glycero-3-phospho-L-serine (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H73N2O10P
CAS Number:
Molecular Weight:
736.96
UNSPSC Code:
12352211
NACRES:
NA.25

form

methanol solution

packaging

pkg of 1 × 1 mL (LM1304-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM1304

concentration

~10 μg/mL (Refer to C of A for lot specific concentration. )

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCC/C=C\CCCC)=O)COC(CCCCCCCCCCCCCCCC)=O.[NH4+]

InChI key

ZBOCVGFQVSWMHT-BPNWTSRKSA-N

General description

Phosphatidylserine (PS) is an acidic phospholipid present in human cerebral cortex. It is abundant in the inner leaflet of the plasma membrane in neural tissues.

Application

17:0-14:1 PS or 1-heptadecanoyl-2-(9Z-tetradecenoyl)-sn-glycero-3-phospho-L-serine has been used as an internal standard:
  • for the detection of lipids by tandem mass spectrometry (MS/MS)
  • to determine lipid concentration in human primary myotubes lipid extracts by mass spectrometry (MS) analysis
  • for phospholipid quantification by MS
  • for lipid extraction by methyl tert-butyl ether (MTBE) method
  • for lipid quantification using liquid chromatography (LC)-MS

Biochem/physiol Actions

Phosphatidylserine (PS) is associated with the Akt, protein kinase C (PKC) and Raf-1 signaling. It supports neuronal survival, neurite growth and synaptogenesis.

Packaging

2 mL Amber Glass Sealed Ampule (LM1304-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hee-Yong Kim et al.
Progress in lipid research, 56, 1-18 (2014-07-06)
Phosphatidylserine (PS) is the major anionic phospholipid class particularly enriched in the inner leaflet of the plasma membrane in neural tissues. PS is synthesized from phosphatidylcholine or phosphatidylethanolamine by exchanging the base head group with serine, and this reaction is
Laurent Perrin et al.
eLife, 7 (2018-04-17)
Circadian regulation of transcriptional processes has a broad impact on cell metabolism. Here, we compared the diurnal transcriptome of human skeletal muscle conducted on serial muscle biopsies in vivo with profiles of human skeletal myotubes synchronized in vitro. More extensive
Ursula Loizides-Mangold et al.
Journal of lipid research, 53(8), 1522-1534 (2012-05-26)
Glycosylphosphatidylinositol (GPI) anchor biosynthesis takes place in the endoplasmic reticulum (ER). After protein attachment, the GPI anchor is transported to the Golgi where it undergoes fatty acid remodeling. The ER exit of GPI-anchored proteins is controlled by glycan remodeling and
Ulrike Bruning et al.
Cell metabolism, 28(6), 866-880 (2018-08-28)
The role of fatty acid synthesis in endothelial cells (ECs) remains incompletely characterized. We report that fatty acid synthase knockdown (FASNKD) in ECs impedes vessel sprouting by reducing proliferation. Endothelial loss of FASN impaired angiogenesis in vivo, while FASN blockade reduced
Alexander Fauland et al.
Journal of lipid research, 52(12), 2314-2322 (2011-10-01)
This work aims to combine chromatographic retention, high mass resolution and accuracy, MS/MS spectra, and a package for automated identification and quantitation of lipid species in one platform for lipidomic analysis. The instrumental setup elaborated comprises reversed-phase HPLC coupled to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service