610009
Avanti
PC Membranes 0.8um
Avanti Research™ - A Croda Brand
Synonym(s):
Extruder PC Membrane, Polycarbonate Membranes 0.8μm 19mm (100/pk)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Avanti Research™ - A Croda Brand
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
Polycarbonate Membrane for Avanti′s Mini-Extruder.
Watch the video to learn more about the Avanti Research™ Mini-Extruder.
Watch the video to learn more about the Avanti Research™ Mini-Extruder.
Biochem/physiol Actions
Polycarbonate membranes are useful to prepare uniform and specific sized liposomes by ordered extrusion. PC Membranes 0.8um is suitable for filtering sarcolemma membrane fragments.
Components
Whatman® Nuclepore Hydrophilic Membrane 10417304
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Whatman is a registered trademark of Cytiva
also commonly purchased with this product
Product No.
Description
Pricing
replacement part
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sarcolemmal Chloride and Potassium Channels from Normal and Myotonic Mouse Muscle Studied in Lipid-Supplemented Vesicles
Biochemical and biophysical research communications, 213(2) (1995)
Preparation of liposomes of defined size distribution by extrusion through polycarbonate membranes
Biochimica et Biophysica Acta - Biomembranes, 557(1), 9-23 (1979)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service