Key Documents
Z220574
Sigma-Aldrich
NMR tube labels
temperature −40-150 °F, pkg of 25 ea
Synonym(s):
Labeling for NMR Tubes, Labels for NMR Tubes, NMR Label Set
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
material
vinyl
packaging
pkg of 25 ea
manufacturer/tradename
Sigma-Aldrich
parameter
−40-150 °F temperature
W
15 mm
Related Categories
General description
Self-laminating, vinyl labels with white, write-on surface resist oil, water, and solvent. Label wraps around 5mm tube for positive sample identification. Supplied with 26 labels on a card, 25 cards per pack.
Choose from one of the most recent versions:
Certificates of Analysis (COA)