Key Documents
Z118028
Luer-to-Tubing Connector
male Luer lock to hose end, for tubing i.d., 1/16 ‑ 3/32 in., nickel plated
Synonym(s):
environmental, veterinary
About This Item
UNSPSC Code:
40183110
NACRES:
NB.03
Pricing and availability is not currently available.
material
nickel plated
feature
for tubing i.d. 1/16 ‑ 3/32 in.
manufacturer/tradename
Cadence Science 6157
General description
Hardware specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. 316 SS fittings are noncontaminating, easy to clean and resistant to corrosion.
Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.
Choose from one of the most recent versions:
Certificates of Analysis (COA)