Merck
SYNC201080
Synthware jacketed reflux condenser
joint: ST/NS 14/10, jacket L 80 mm
Synonym(s):
Synthware C201080
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.51
UNSPSC Code:
41121800
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
joint
joint: ST/NS14/10
packaging
pack of 1 ea
manufacturer/tradename
C201080
jacket L
80 mm
General description
Synthware reflux condenser features a 80 mm jacketed length for superior cooling efficiency. I.D. of inner tube is designed large enough to accept a thermometer. Supplied with one holed compression cap and one Viton® O-ring. Made from durable borosilicate glass.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Global Trade Item Number
SKUGTIN
SYNC201080-1EA04061832190808