Skip to Content
Merck
All Photos(1)

Documents

P49902

Sigma-Aldrich

Pregnenolone acetate

99%

Synonym(s):

3β-Acetoxy-5-pregnen-20-one, 5-Pregnen-3β-ol-20-one acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H34O3
CAS Number:
Molecular Weight:
358.51
Beilstein:
2062850
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder

optical activity

[α]20/D +19°, c = 1 in ethanol

mp

149-152 °C (lit.)

SMILES string

CC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC[C@@H]4C(C)=O)[C@@H]3CC=C2C1

InChI

1S/C23H34O3/c1-14(24)19-7-8-20-18-6-5-16-13-17(26-15(2)25)9-11-22(16,3)21(18)10-12-23(19,20)4/h5,17-21H,6-13H2,1-4H3/t17-,18-,19+,20-,21-,22-,23+/m0/s1

InChI key

CRRKVZVYZQXICQ-RJJCNJEVSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A McMaster et al.
The Journal of endocrinology, 197(2), 205-211 (2008-04-25)
There is increasing evidence that temporal factors are important in allowing cells to gain additional information from external factors, such as hormones and cytokines. We sought to discover how cell responses to glucocorticoids develop over time, and how the response
Anam Ansari et al.
Steroids, 148, 99-113 (2019-05-15)
A series of steroidal thiazolidinone derivatives have been synthesized through one-pot multicomponent reaction involving steroidal ketone, thiosemicarbazide/methyl-thiosemicarbazide and DMAD in presence of AlCl3 as a Lewis acid catalyst. Among all the synthesized steroidal thiazolidinone derivatives, compound 7-9 (ST 7-9) were
Xuelu Ding et al.
Rapid communications in mass spectrometry : RCM, 32(16), 1439-1450 (2018-05-29)
Neurosteroids are important signalling molecules that modulate neuronal activity. Their low concentrations and low volatility make neurosteroid detection and quantification by ambient mass spectrometry challenging. Here we develop a reactive low-temperature plasma mass spectrometry (LTP-MS) method and demonstrate its potential
Jamie L York et al.
Food chemistry, 326, 126939-126939 (2020-05-16)
Hen egg yolk from chicken eggs were examined for their mycotoxin, hormone, and fat-soluble vitamin content. A method was developed for multi-class analysis of egg yolk using a dilute/precipitate, centrifuge, and shoot process coupled with on-line sample clean-up using restricted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service