MP12002010
MakroPor, silicon membrane
thickness 50 μm, pore diameter 2.5 μm, pore size 4.2 μm (interpore distance), size 20 mm × 20 mm, pkg of 10 pieces
About This Item
Recommended Products
material
silicon membrane
description
FZ single crystal Si
Pore geometry: trigonal arrangement
packaging
pkg of 10 pieces
size
20 mm × 20 mm
thickness
50 μm
pore size
2.5 μm pore diameter
4.2 μm (interpore distance)
compatibility
for use with applications requiring mechanical and chemical stability with high order of the pores
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service