JRD1724
5-Amino-1-(2-fluorophenyl)-1H-pyrazole-4-carbohydrazide
AldrichCPR
Synonym(s):
5-Amino-1-(2-fluorophenyl)-1H-pyrazole-4-carboxylic acid hydrazide
About This Item
Recommended Products
functional group
amine
fluoro
hydrazine
SMILES string
NC(N(C(C=CC=C1)=C1F)N=C2)=C2C(NN)=O
FC1=CC=CC=C1N2C(N)=C(C(O)=NN)C=N2
InChI
1S/C10H10FN5O/c11-7-3-1-2-4-8(7)16-9(12)6(5-14-16)10(17)15-13/h1-5H,12-13H2,(H,15,17)
InChI key
ZYBIAUHQEMAAKF-UHFFFAOYSA-N
Other Notes
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service