Skip to Content
Merck
All Photos(1)

Key Documents

GF59595047

Hastelloy® C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn

100mm tube, 9.5mm outside diameter, 0.9mm wall thickness, 7.7mm inside diameter

Synonym(s):

NI147300

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 595-950-47

L × wall thickness

100 mm × 0.9 mm

O.D. × I.D.

9.5 mm × 7.7 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Hastelloy is a registered trademark of Haynes International, Inc.
Hastelloy is a registered trademark of Haynes International, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service