Key Documents
DWK45066C-13
KIMBLE® Solid Cap
black phenolic cap (with PTFE Liner), cap for 13-415
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
material
PTFE/rubber liner
black phenolic cap (with PTFE Liner)
sterility
non-sterile
feature
class B
packaging
case of 300 ea
manufacturer/tradename
Kimble® 45066C-13
fitting
cap for 13-415
General description
Class B Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
  • Specially formulated phenolic cap material
  • Autoclavable
  • White rubber liners
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class B requirement
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)