Skip to Content
Merck
All Photos(1)

Key Documents

D15901

Sigma-Aldrich

5,6-Diamino-1,3-dimethyluracil hydrate

technical grade

Synonym(s):

4,5-Diamino-1,3-dimethyl-2,6-dioxotetrahydropyrimidine, 5,6-Diamino-1,3-dimethyl-2,4(1H,3H)-pyrimidinedione, NSC 15493

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N4O2 · xH2O
CAS Number:
Molecular Weight:
170.17 (anhydrous basis)
Beilstein:
157451
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

solid

mp

210-214 °C (dec.) (lit.)

SMILES string

O.CN1C(N)=C(N)C(=O)N(C)C1=O

InChI

1S/C6H10N4O2.H2O/c1-9-4(8)3(7)5(11)10(2)6(9)12;/h7-8H2,1-2H3;1H2

InChI key

FNLDCZCNGSQKIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in the synthesis of biologically active molecules including:
  • 1H-Imidazol-1-yl substituted 8-phenylxanthines for use as adenosine receptor ligands
  • Pyrimidinyl purinediones via cyclocondensation
  • Theophylline derivatives as adenosine receptor antagonists
  • Pteridine analogs for use as monoamine oxidase B and nitric oxide synthase inhibitors
  • Fused tricyclic xanthines
  • Cadmium uraciliminoethylpyridine complexes for use as antiproliferatives against C6 glioma cells

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service