Key Documents
CLS7095BNMR
Corning® Pasteur pipettes, non-sterile
L 13 1/4 in. (337 mm), long tip, soda lime
Synonym(s):
Pipettes, pipettes, serological pipets, serological pipettes
About This Item
UNSPSC Code:
41121500
NACRES:
NB.04
material
borosilicate glass
sterility
non-sterile
feature
graduations: no
subdivision: no
packaging
case of 100 ea
pack of 100 ea
manufacturer/tradename
Corning 7095B-NMR
L
13 1/4 in. (337 mm)
General description
  • Corning® disposable Pasteur pipettes are made of sturdy soda-lime glass.
  • Single-use pipettes prevent the risk of cross-contamination of valuable specimens or laboratory reagents.
  • Feature a constriction one inch below the top to facilitate plugging.
  • Ideal for use in non-volumetric transfer work in bacteriology, immunology, hematology, and serology studies, as well as blood bank and clinical chemistry procedures.
Disposable. Features a constriction one inch below the top to facilitate plugging. Ideal for rapid nonvolumetric work in bacteriology, immunology, hematology, serology studies, blood bank and clinical chemistry procedures.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)