CLS4985125
Pyrex® narrow-mouth Erlenmeyer flask with screw-cap
capacity 125 mL
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 4985
About This Item
Recommended Products
material
borosilicate glass
narrow-mouth
phenolic cap
screw top cap
neck thread size
24-410
sterility
non-sterile
feature
cap
closure type Screw cap
subdivision 25 mL
packaging
pack of 12
case of 24
manufacturer/tradename
Corning 4985-125
volume measuring range
50-125 mL
capacity
125 mL
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service