CLS2679425
Corning® polypropylene closure
orange
Synonym(s):
Corning closure
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.43
Recommended Products
description
fits neck O.D. 25 mm
manufacturer/tradename
Corning 26794-25
color
orange
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For culture tubes and flasks with long rimless necks. Position closure open for gas exhange or closed for humidified environment. Internal drip ring minimizes contamination.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service