BR13903
BRAND® compact SILBERBRAND burette, with PTFE stopcock
volume 10 mL
Synonym(s):
Burettes, Compact burettes, SILBERBRAND
All Photos(3)
About This Item
Recommended Products
material
glass
manufacturer/tradename
BRAND 13903
volume measuring range
10 mL, accuracy: ±0.05 mL
volume
10 mL
Related Categories
General description
The New Generation of Burettes from BRAND. With PTFE stopcock, fast to dismantle and easy to clean. Simple repair - all individual components replaceable. Boro 3.3, removable PTFE stopcock. Error limits according to class B, DIN EN ISO 385. Calibrated to deliver (TD, Ex).
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)