Skip to Content
Merck
All Photos(1)

Documents

938874

Sigma-Aldrich

NanoFabTx PEG-PEI

new

for gene delivery

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352142

Quality Level

application(s)

advanced drug delivery

storage temp.

2-8°C

General description

The NanoFabTxPEG-PEI for gene delivery has been developed for the synthesis of PEG-PEI polyplex nanoparticles for use in oligonucleotide, nucleic acid, and other gene delivery applications. The PEG-PEI cationic copolymer combines the unique properties of both polyethylene glycol (PEG) and polyethylenimine (PEI), resulting in a versatile and efficient delivery platform. The PEG component provides excellent biocompatibility and stability, while the branched PEI component offers superior drug encapsulation and controlled release capabilities.

Application

With the NanoFabTx(TM) PEG-PEI, researchers can simplify their drug development workflow. The ready to use polymer enables the screening of different drug modalities such as oligonucleotides, nucleic acids (mRNA, DNA, siRNA), and vaccines without the need to develop new drug delivery formulations from scratch, ultimately speeding up the time it takes for innovative therapeutics to go from the bench to the clinic. PEG-coupled PEI has been used in many drug delivery applications and has been found to improve the stability, prevent aggregation, decrease interactions with serum proteins thus improving circulation times, and enhance gene transfection. Here, the NanoFabTx PEG-PEI has been optimized to suit your gene delivery needs by offering superior mRNA complexation, polyplex size, and cell viability. A step by step protocol is can be located under the “More Documents” section of this page and provide detailed guidance on how to use the NanoFabTx PEG-PEI for gene delivery.

Features and Benefits

  • A PEG-PEI polymer curated and tested for gene delivery
  • The NanoFabTxPEG-PEI can be used to create polyplex nanoparticles with a variety of nucleic acids and other drug modalities
  • The step-by-step protocol provides detailed instructions on synthesizing and characterizing mRNA polyplexes
  • Reproducible polyplexes can be synthesized in a variety of sizes between 70nm-150nm by altering key parameters such as N/P ratio
  • The nucleic acid-encapsulated polyplex nanoparticles created using the NanoFabTx PEG-PEI result in high in vitro transfection efficiency and high cell viability in HeLa and HEK293 cell lines
For more information, please refer to the protocol under the document section of this page.

Legal Information

NANOFABTX is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delivery of messenger RNA using poly(ethylene imine)?poly(ethylene glycol)-copolymer blends for polyplex formation: Biophysical characterization and in vitro transfection properties
Debus H, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 148, 334-343 (2010)
Jing Wang et al.
Biomaterials science, 4(9), 1351-1360 (2016-07-19)
Zwitterionic poly(carboxybetaine) (PCB), poly(2-methacryloyloxyethyl phosphorylcholine) (PMPC) and non-ionic poly(ethylene glycol) (PEG), which have similar degrees of polymerization, were grafted to branched polyethyleneimine (PEI) to generate PCB-grafted PEI (PEI-PCB), PMPC-grafted PEI (PEI-PMPC) and PEG-grafted PEI (PEI-PEG) copolymers, respectively. These grafted PEI
Chunxi Liu et al.
Biomaterials, 34(10), 2547-2564 (2013-01-22)
Co-delivery of nucleic acids and chemotherapeutics has a potential to efficaciously treat human diseases via their synergetic effects. Activable therapeutic tools at the nanoscale are suitable platforms for combination therapy. In this study, we have developed a multifunctional nanoscaled delivery

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service