Skip to Content
Merck

929921

Sigma-Aldrich

Allochenodeoxycholic acid-2,2,4,4-d4 solution

≥98 atom % D, ≥98% (CP), 100 μg/mL in methanol

Synonym(s):

(R)-4-((3R,5R,7R,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl-2,2,4,4-d4)pentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24D4H36O4
Molecular Weight:
396.60
UNSPSC Code:
12142200
NACRES:
NA.13

Pricing and availability is not currently available.

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

concentration

100 μg/mL in methanol

mass shift

M+4

SMILES string

O[C@@H]1C([2H])([2H])C[C@@]2(C)[C@](C[C@@H](O)[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CCC(O)=O)([H])C1([2H])[2H]

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
929948929964SMB00445
mass shift

M+4

mass shift

M+3

mass shift

M+5

mass shift

-

isotopic purity

≥98 atom % D

isotopic purity

≥98 atom % 13C

isotopic purity

≥98 atom % 13C

isotopic purity

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

concentration

100 μg/mL in methanol

concentration

100 μg/mL in methanol

concentration

-

concentration

-

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service