924733
Cross-shaped channel chip - 50 μm channel (double T-junction)
Fluidic 201, Cyclic Olefin Polymer (COP) with Luer Interface
Synonym(s):
Microfluidic
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
UNSPSC Code:
42142600
NACRES:
NA.25
Application
This is a Cross-shaped channel chip with an extended size platform Ι and a Luer fluidic interface.
Cross-shaped channel chip - 50 μm channel (double T-junction), Fluidic 201, Cyclic Olefin Polymer (COP) with Luer Interface.
Cross-shaped channel chip - 50 μm channel (double T-junction), Fluidic 201, Cyclic Olefin Polymer (COP) with Luer Interface.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Dongfei Liu et al.
Lab on a chip, 17(11), 1856-1883 (2017-05-10)
The microfluidic technique has brought unique opportunities toward the full control over the production processes for drug delivery carriers, owing to the miniaturisation of the fluidic environment. In comparison to the conventional batch methods, the microfluidic setup provides a range
Sharma T Sanjay et al.
Advanced drug delivery reviews, 128, 3-28 (2017-09-19)
Conventional systematically-administered drugs distribute evenly throughout the body, get degraded and excreted rapidly while crossing many biological barriers, leaving minimum amounts of the drugs at pathological sites. Controlled drug delivery aims to deliver drugs to the target sites at desired
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service