Skip to Content
Merck
All Photos(1)

Key Documents

924415

Sigma-Aldrich

Sodium 2-methyl-4,5-dihydro-1,3-thiazole-4-carboxylate

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6NNaO2S
CAS Number:
Molecular Weight:
167.16
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.21

Assay

≥95%

form

powder

storage temp.

2-8°C

SMILES string

O=C(C(CS1)N=C1C)O[Na]

InChI

1S/C5H7NO2S.Na/c1-3-6-4(2-9-3)5(7)8;/h4H,2H2,1H3,(H,7,8);/q;+1/p-1

InChI key

KDIYEJLRFZLXKU-UHFFFAOYSA-M

General description

Sodium 2-methyl-4,5-dihydro-1,3-thiazole-4-carboxylate is a building block that can be used in the synthesis of Methylthiazolinyl-S-NAC in combination with other reagents to produce thioester-linked product. It is often used in the chemical synthesis of specific enzyme substrates and standards for biochemical research and analysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidase domains in epothilone and bleomycin biosynthesis: thiazoline to thiazole oxidation during chain elongation
Schneider TL, et al.
Biochemistry, 42, 9722-9730 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service