Skip to Content
Merck
All Photos(1)

Key Documents

909653

Sigma-Aldrich

Stable Isotope Labeled Amino Acid Mixture for Mass Spec

pkg of 25 nmol (lyophilized powder)

Synonym(s):

Amino acid mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352209
NACRES:
NA.12
Pricing and availability is not currently available.

form

powder

Quality Level

packaging

pkg of 25 nmol (lyophilized powder)

technique(s)

mass spectrometry (MS): suitable

storage temp.

−20°C

Related Categories

General description

Each tube contains a lyophilized mixture of approximately 25 nmoles of each amino acid. Refer to the batch-specific Certificate of Analysis for exact quantities.

L-Alanine-13C3,15N
L-Arginine-13C6,15N4 hydrochloride
L-Asparagine-15N2
L-Aspartic acid-13C4,15N
DL-Cystine-3,3,3′,3′-d4
L-Glutamic acid-13C5,15N
L-Glutamine-15N2
Glycine-13C2,15N
L-Histidine-15N3
L-Isoleucine-13C6,15N
L-Leucine-13C6,15N
L-Lysine-13C6,15N2 hydrochloride
L-Methionine-15N
L-Phenylalanine-13C9,15N
L-Proline-13C5,15N
L-Serine-13C3,15N
L-Threonine-13C4,15N
L-Tryptophan-15N2
L-Tyroxine-13C9,15N
L-Valine-13C5,15N

Application

Used as mass spec standard for determination of amino acids in sample.
Refer to attached protocol for recommended use.
Please see attachment for information concerning precursor and product ions.
Product has been designed to minimize potential signal overlap between sample metabolites and SIL amino acids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

  1. What is the concentration?

    1 answer
    1. Each tube contains a lyophilized mixture of approximately 25 nmoles of each amino acid. Refer to the batch-specific Certificate of Analysis for exact quantities.

      L-Alanine-13C3,15N
      L-Arginine-13C6,15N4 hydrochloride
      L-Asparagine-15N2
      L-Aspartic acid-13C4,15N
      DL-Cystine-3,3,3',3'-d4
      L-Glutamic acid-13C5,15N
      L-Glutamine-15N2
      Glycine-13C2,15N
      L-Histidine-15N3
      L-Isoleucine-13C6,15N
      L-Leucine-13C6,15N
      L-Lysine-13C6,15N2 hydrochloride
      L-Methionine-15N
      L-Phenylalanine-13C9,15N
      L-Proline-13C5,15N
      L-Serine-13C3,15N
      L-Threonine-13C4,15N
      L-Tryptophan-15N2
      L-Tyroxine-13C9,15N
      L-Valine-13C5,15N

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service