88755
(R)-1-[(SP)-2-(Di-tert-butylphosphino)ferrocenyl]ethylbis(2-methylphenyl)phosphine
97%
Synonym(s):
(1R)-1-[Bis(1,1-dimethylethyl)phosphino]-2-[(1R)-1-[bis(2-methylphenyl)phosphino]ethyl]ferrocene (acc to CAS), Josiphos SL-J505-1
About This Item
Recommended Products
Quality Level
Assay
97%
form
powder
optical purity
ee: ≥99%
functional group
phosphine
SMILES string
[Fe].[CH]1[CH][CH][CH][CH]1.C[C@H]([C]2[CH][CH][CH][C]2P(C(C)(C)C)C(C)(C)C)P(c3ccccc3C)c4ccccc4C
InChI
1S/C29H39P2.C5H5.Fe/c1-21-15-10-12-18-25(21)30(26-19-13-11-16-22(26)2)23(3)24-17-14-20-27(24)31(28(4,5)6)29(7,8)9;1-2-4-5-3-1;/h10-20,23H,1-9H3;1-5H;/t23-;;/m1../s1
InChI key
DSZYQALCGVODLB-MQWQBNKOSA-N
General description
Application
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service