Skip to Content
Merck
All Photos(2)

Key Documents

81840

Sigma-Aldrich

Dimethyl propargylmalonate

≥95.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CCH2CH(COOCH3)2
CAS Number:
Molecular Weight:
170.16
Beilstein:
3539408
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.444

bp

93-95 °C/7 mmHg (lit.)

density

1.119 g/mL at 20 °C (lit.)

functional group

ester

SMILES string

COC(=O)C(CC#C)C(=O)OC

InChI

1S/C8H10O4/c1-4-5-6(7(9)11-2)8(10)12-3/h1,6H,5H2,2-3H3

InChI key

PWQAXFWWMXTVFT-UHFFFAOYSA-N

Application

Dimethyl propargylmalonate can be used as a reactant to synthesize:
  • Nitro methylenecyclopentanes by [3+2] annulation reaction with various nitroalkenes in the presence of Triton B.
  • Propargylmalonamides intermediates, applicable in the preparation of ″click BOX″ ligands by copper-catalyzed cycloaddition and oxazoline ring formation reaction.
  • Cyclopentene derivatives by reacting with various α, β-unsaturated ketones using a combination of organocatalysts and transition metal catalysts.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of ?Click BOX? ligands and preliminary results on their application in the asymmetric copper catalyzed Henry reaction of o-methoxybenzaldehyde
Giunta D, et al.
Results in Chemistry, 3, 100122-100122 (2021)
Combination iminium, enamine and copper (I) cascade catalysis: a carboannulation for the synthesis of cyclopentenes
Yang T, et al.
Chemical Communications (Cambridge, England), (25), 2923-2925 (2008)
One-pot Michael addition/intramolecular carbocyclization of dimethyl propargylmalonate with nitroalkenes: A new stereoselective [3+ 2] annulation to 1-nitro 2-methylenecyclopentanes
Guillaume M, et al.
Synlett, 2002(11), 1883-1885 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service